VWR® Dual-Traceable, Thermometers
Supplier: VWR Collection
620-2689EA 86.5 EUR
620-2689 620-2688
VWR® Dual-Traceable, Thermometers
Thermometers Digital Thermometers
These dual-probe thermometers give accurate readings of two separate areas in refrigerators/freezers. Allows putting one probe in a freezer and another in a refrigerator (or measure two areas of a refrigerator).
  • Two-part display shows °F or °C probe temperatures in distinct sections of refrigerator or freezer
  • Easy to operate, always ready unit quickly recalls exact date and time when MIN/MAX temperatures occur
  • Audible and visual alarm feature provides alert when temperature rises above or falls below two set points, alarm is programmable in 1 degree