Twin-frosted adhesion slides, HistoBond®
Supplier: MARIENFELD
0810000 0810001 0830001
631-0624EA 16.9 EUR
631-0624 631-0625 630-2830
Twin-frosted adhesion slides, HistoBond®
Slides Adhesive Slides
The glass surface of the HistoBond® slide is loaded with a permanent positive charge, which fastens all types of section to the glass.
  • Ground edges 90°, twin-frosted on both sides
  • Silane-treated, pre-cleaned and ready for use
  • Autoclavable with silky frosted marking area of 20 mm at one end on both sides
In most cases, this reliable bonding makes it possible to dispense with double experiments which saves time in section preparations.