Greenough Stereo Microscopes, Ivesta 3
Supplier: LEICA MICROSYSTEMS
10450961 10450981 10450982 10450983 12730601 12730602 10450981_P 10450982_P 10450983_P
630-3534EA 385 EUR
630-3534 630-3527 630-3531 630-3530 630-3533 630-3535 630-3538 630-3537 630-3536
Greenough Stereo Microscopes, Ivesta 3
Microscopes
Ivesta 3 – For inspection and rework only, but no camera for documentation is needed.
Ivesta 3 with C-mount – Gives you the flexibility to add a camera if needed and thus turn it into a digital microscope solution.
Ivesta 3 with integrated camera – For digital image sharing directly on different devices, such as, a monitor, mobile device, or computer.
  • Save time refocusing as you can see every important detail at a glance
  • FusionOptics allows you to perceive your samples in 3D with up to 3× larger depth of field
  • Change from sample overview to detail easily