VWR® VisiScope® IT417 PH Inverted Microscopes
Supplier: VWR Collection
630-3548EA 3320 EUR
630-3548 630-3549
VWR® VisiScope® IT417 PH Inverted Microscopes
Microscopes
This brightfield, phase contrast microscope is the ideal solution for fast and reliable routine inspections, especially in in vivo cell culture applications.
  • High-efficiency LED illumination with brightness control: Ideal for all transmitted light observations
  • Condenser can be removed to extend the working distance up to 150 mm
  • Fixed specimen stage for fatigue-free operation
  • Two-stage insert (glass and metal) with hole for small dimension specimens
  • Plan achromatic LWD objectives (IOS) with anti-fungus treatment
Innovative design, interchangeableMore Product Information