NANOCOLOR® Photometric Standard Test Kits for Iron
Supplier: MACHEREY-NAGEL
91836 918128
634-8129EA 119 EUR
634-8129 MANA918128
NANOCOLOR® Photometric Standard Test Kits for Iron
Environmental Test Kits
Photometric test kits for the determination of iron in water samples.
  • Ready-to-use reagents for up to 500 determinations
  • Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
  • Low costs per determination
  • Easy dilutions for analysis of higher concentrations
The test kits are evaluated in 10 or 50 mm rectangular cuvettes. They are preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.
Remark: Measuring range on NANOCOLOR VIS II
Delivery information: Rugged box with reagents for 250 tests.