Pipette stand
Supplier: Socorex Isba
320.340
635-0362EA 99.4 EUR
635-0362
Pipette stand
Pipette Stands
Pipette stand made of light grey polyamide.
  • For 3 single and/or multi channel pipettes
  • Innovative design fits all Calibra® and Acura® models and allows easy reach of each instrument
  • Heavy bottom plate and anti-slip pads guarantee stability