Anti-CD8A Rat Monoclonal Antibody (Peridinin Chlorophyll/Cy5.5®) [clone: 2.43]

Supplier: Tonbo Biosciences
65-1886-U100 65-1886-U025
65-1886-U100EA 226 EUR
65-1886-U100 65-1886-U025
Anti-CD8A Rat Monoclonal Antibody (Peridinin Chlorophyll/Cy5.5®) [clone: 2.43]
Antibodies
The 2.43 antibody reacts with the 32 to 34 kDa alpha subunit of mouse CD8, known as CD8a or CD8 alpha. CD8a can form a homodimer (CD8 alpha-alpha), but is more commonly expressed as a heterodimer with a second chain known as CD8b or CD8 beta. CD8 acts as a co-receptor in antigen recognition and subsequent T cell activation induced by binding of the T cell receptor (TCR) to antigen-bearing MHC Class I molecules. The cytoplasmic domains of CD8 provide binding sites for the tyrosine kinase lck and facilitate intracellular signaling events that lead to T cell activation, development, andMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol CD8A
Environmentally Preferable
Antigen name CD8 antigen, alpha chain
Antigen synonyms p32|Lyt-66|BB154331|Ly-2|Leu2|Ly-35|MAL|CD8|Ly-B
Antibody type Primary
Clonality Monoclonal
Conjugation PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Clone 2.43
Reactivity Mouse
Host Rat
Gene ID 12525
Isotype IgG2b
ImmunoChemistry Yes
Flow cytometry Yes
Neutralisation Yes
Purification Yes
Cross adsorption No
Storage temperature Cool
Tested applications Flow Cytometry

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR