pH indicator papers, universal/special
Supplier: MACHEREY-NAGEL
90201 90202 90203 90205 90206 90207 90208 90209 90210 90211 90212 90213 90225 90226 90227 90228 90229 90230 90231 90233 90224 90204 90214 90232 90234
662-0209EA 12.7 EUR
662-0209 662-0230 662-1866 662-0231 MANA90206 662-0206 MANA90208 662-0232 662-0233 MANA90211 MANA90212 MANA90213 MANA90225 MANA90226 MANA90227 MANA90228 MANA90229 MANA90230 MANA90231 MANA90233 662-0193 662-0204 MANA90214 MANA90232 MANA90234
pH indicator papers, universal/special
Test Papers pH Indicator Papers and Test Strips
pH indicator papers have been available for decades and are the standard for many applications. For each pH value these papers show a single colour which can be matched with the colour scale at intervals of 0,2 to 1 pH units.
  • Long term stable and protected against external influences due to plastic reel
  • No accessories needed - always ready to use
  • Reliable results due to easy and accurate read-off from colour scale
  • Indicator papers are supplied as rolls with 5 m length and 7 mm width
pH indicators are manufactured from high quality filter papers. TheMore Product Information