pH/mV/°C meters, bench, inoLab® pH 7110 / 7310
Supplier: WTW
1AA110 1AA112 1AA114 1AA120 1AA310 1AA312 1AA314 1AA320 1AA310P 1AA320P
662-1357EA 801 EUR
662-1357 662-1349 662-1358 662-1359 662-1362 662-1364 662-1366 662-1368 662-1363 662-1369
pH/mV/°C meters, bench, inoLab® pH 7110 / 7310
Electrochemistry Meters pH and Multi-Parameter Meters
High quality instruments with advanced measuring technology, offering functions including automatic AutoRead, CMC (Continuous Measurement Control) and QSC (Quality Sensor Control) making measurements more convenient and reliable.
inoLab® 7110: Accurate measurements
The inoLab® 7110 series is ideal for routine measurements in general laboratory applications. Whether measuring pH in environmental, chemical, pharmaceutical, medical or food and beverage industries, this series offers advanced features in a simple to use package.
- AutoRead function ensures repeatable results
- EasyMore Product Information