pH/mV/°C/ion meter, bench, SevenExcellence™, S500
Supplier: Mettler - Toledo
30046248 30046250 30046249 30046251
662-1478EA 2930 EUR
662-1478 662-1480 662-1479 662-1483
pH/mV/°C/ion meter, bench, SevenExcellence™, S500
Electrochemistry Meters pH and Multi-Parameter Meters
This single-channel instrument can easily handle the most complex applications and meets stringent requirements in regulated markets. The meter is flexible and can be modularly expanded with additional measurement parameters at any time. Special methods for the most common incremental ion measurement techniques are included, such as standard addition, subtraction and sample addition and subtraction methods.
  • Methods for incremental ion measurements, guiding the user through the individual steps
  • Full ion support: 12 most commonly used ion types are pre-