Dissolved oxygen meters, handheld, Seven2Go™ S4 / S9 Pro
Supplier: Mettler - Toledo
30207939 30207971 30207970 30207960 30207959 30207972 30207958
662-1849EA 1890 EUR
662-1849 662-1857 662-1856 662-1852 662-1851 662-1858 662-1850
Dissolved oxygen meters, handheld, Seven2Go™ S4 / S9 Pro
Electrochemistry Meters Oxygen Meters
These robust meters with polarographic or optical sensor provide durability and one-handed operation, making them ideally suited for mobile applications in the laboratory, at-line, or outdoors. Thanks to their IP 67 protection and drop test resistance, the devices are able to withstand harsh and demanding environments.
  • New intuitive menu guarantees ease of use
  • T-Pad with smart button arrangement enables easy and fast navigation
  • Instrument automatically recognises InLab ISM sensors and stores sensor ID, serial number, and last calibration data
  • Automatic or