pH/mV/°C meters, bench, FiveEasy™ F20 / FiveEasy™ Plus FP20
Supplier: Mettler - Toledo
30266658 30266626 30266627 30266628 30266629 30266940 30266941
662-2011EA 584 EUR
662-2011 662-2001 662-2002 662-2003 662-2004 662-2005 662-2006
pH/mV/°C meters, bench, FiveEasy™ F20 / FiveEasy™ Plus FP20
Electrochemistry Meters pH and Multi-Parameter Meters
These bench top instruments provide high quality pH/mV measurements with the simple click of a button. Thanks to their intuitive operation, sleek design and simple data export functions they are easy to set up and to use. Ideal for use in food and beverage processing, agriculture, environment, industry and for education purposes.
  • Robust ABS housing and large, well-structured display for easy viewing
  • Compact design: Requires very little bench space in the lab
  • Sensor holder can be easily removed and stored on the side of the meter after use
  • Acoustic and