pH electrodes, InLab® ISM
Supplier: Mettler - Toledo
51344102 51344103 51344104 51344055 51344155 51344211 51344072 51344172 51343156 30014096 30247850 30248830 30248832
662-1153EA 493 EUR
662-1153 662-1750 662-1910 662-1234 662-1236 662-1239 662-1235 662-1238 662-1245 663-0198 662-2032 662-2037 662-2038
pH electrodes, InLab® ISM
Electrodes pH Electrodes
High performance electrodes combining robustness with state-of-the-art measurement technology. Fully traceable.
  • Calibration data and Sensor ID are automatically transferred to the meter
  • New calibration data are stored in the sensor
  • Back-up certificate guaranteed: Initial factory calibration is stored in the sensor
  • Conclusive calibration history: The last five calibrations are stored in the sensor
  • Easy lifetime monitoring: The maximum temperature that the sensor has been exposed to is monitored automatically
The revolutionary Intelligent SensorMore Product Information