Orion™ Versa Star Pro™ pH/mV/ISE Meters, Bench
Supplier: Thermo Orion
VSTAR40A VSTAR40A2 VSTAR40B2 VSTAR40B
662-2306EA 1980 EUR
662-2306 662-2307 662-2309 662-2308
Orion™ Versa Star Pro™ pH/mV/ISE Meters, Bench
Electrochemistry Meters pH and Multi-Parameter Meters
These bench top meters with conductivity module meet most challenging applications for pH, ion concentration, mV, ORP and temperature. The meters offer interchangeable measurement modules that allow multiple users to customise four separate channels to meet their specific requirements.
  • Colour display with adjustable text sizes; numeric keypad; onscreen measurement stability indicator
  • Auto-read to hold a stable reading; ten password-protected methods per channel
  • Audible alarms automatically notify users when a set offset value, high/low limit value, and