Combined titration electrodes, plug and play
Supplier: Mettler - Toledo
51109520 51109530
662-2476EA 550 EUR
662-2476 662-2477
Combined titration electrodes, plug and play
Electrodes pH Electrodes
Plug & Play combined silver ring electrode for argentometric titrations.
  • Plug & Play sensor with integrated sensor chip for secure handling
  • Sensor ship stores data such as sensor type, serial number, calibration data and usable life, which is automatically read out to the titrator setup when connected
  • Ceramic frit junction
  • Pure silver sensing element
  • ARGENTHAL™ reference system
  • Refillable reference electrolyte (1 mol/l KNO3)
Typical applications include: Titrations of halides, sulfides, hydrogen sulfide, mercaptans and cyanides in aqueousMore Product Information