Conductivity sensors, InLab® Trace
Supplier: Mettler - Toledo
30014097 30014099
663-0264EA 1590 EUR
663-0264 663-0265
Conductivity sensors, InLab® Trace
Electrodes Conductivity Sensors
This digital conductivity sensor has a high precision temperature probe. The conductivity and temperature signals are digitised and combined through a special algorithm for unmatched conductivity accuracy.
  • Optional PP flow cell provides protection from carbon dioxide: Ensures very high repeatability and reproducibility
  • Flow cell was built at an angle to prevent air bubbles being trapped
  • Special temperature compensation mode to increase the reliability of readings
  • Certified cell constant makes calibration superfluous
Ideal for low conductivityMore Product Information