Conductivity meters, handheld, Cond 3110 / 3310
Supplier: WTW
2CA100 2CA101 2CA103 2CA300 2CA301
663-8121EA 743 EUR
663-8121 663-8122 663-8124 663-8129 663-8130
Conductivity meters, handheld, Cond 3110 / 3310
Electrochemistry Meters Conductivity Meters
Waterproof handheld meters designed for unlimited outside use.
  • Large display allows excellent readability even under bad lighting conditions
  • Robust housing and sealed silicone keypad with protection class IP 66/67
Cond 3110: Reliable, clearly designed meter for basic and standard measurements, e.g. in educational applications.
- Large LCD
- Tactile response offering clear operation
- Built-in timer ensures accurate measurements
- Automatic temperature compensation
- Up to 1000 hours of continuous operation time with one battery set