Dissolved oxygen meter, handheld, Orion Star™ A123
Supplier: Thermo Orion
STARA1230 STARA1235
664-0143EA 669 EUR
664-0143 664-0144
Dissolved oxygen meter, handheld, Orion Star™ A123
Electrochemistry Meters Oxygen Meters
The Orion Star™ A123 dissolved oxygen handheld meter offers easy operation for basic analysis in the field. Ideal for taking routine dissolved oxygen and temperature measurements.
  • Measure dissolved oxygen in percent saturation or mg/L with temperature using polarographic DO probes
  • Calibrate using water-saturated air or custom value (Winkler titration)
  • Automatically compensate DO readings using ATC temperature, manual salinity input and manual barometric pressure
  • Simplified, easy-to-read display shows main measurement with temperature, stability indicator