Multi-parameter meters, handheld, SevenGo Duo™ / Duo pro™ (SG23 / SG78)
Supplier: Mettler - Toledo
51302600 51302601 51302602 51302603 51302620 51302621 51302622 51302623 51302625
665-0200EA 1090 EUR
665-0200 665-0191 665-0192 664-0163 665-0201 665-0195 665-0196 664-0162 664-0161
Multi-parameter meters, handheld, SevenGo Duo™ / Duo pro™ (SG23 / SG78)
Electrochemistry Meters pH and Multi-Parameter Meters
Portable dual channel meters for the simultaneous determination of pH, mV, rel. mV, ion (SG78), conductivity, TDS (total dissolved solids), salinity or specific resistance.
SevenGo Duo™ - pH/conductivity meter (IP 67) for routine use
- High contrast display with large digits, rugged watertight housing (IP 67)
- Automatic adjustment of cell constant
- 3-point pH calibration with selection from four pre-defined and one user-defined buffer group
- Data memory for 99 data records
- Automatic end point detection, automatic buffer recognition and temperature compensation
SevenGo Duo pro™ -More Product Information