pH/mV/ISE/°C meters, bench top, Orion Star™ A214
Supplier: Thermo Orion
STARA2140 STARA2145 STARA2146 STARA2147 STARA2148
665-0269EA 1390 EUR
665-0269 665-0270 665-0477 665-0478 665-0479
pH/mV/ISE/°C meters, bench top, Orion Star™ A214
Electrochemistry Meters pH and Multi-Parameter Meters
These bench top meters record accurate and reliable pH, ion concentration, mV, ORP and temperature measurements. They are ideal for a wide range of applications and advanced pH or ion analysis in the lab.
  • Up to a five point pH calibration using automatic buffer recognition of NIST or DIN buffer groups
  • Up to a five point ISE (Ion Selective Electrode) calibration with selectable units of ppm, M, mg/L, %, ppb or none
  • Advanced ISE features include non-linear selectable auto-blank and low range stability
  • Measurement stability indicator and selectable read modes