Anti-KLRB1 Mouse Monoclonal Antibody [clone: HP-3G10]

Supplier: Tonbo Biosciences
70-1619-U100
70-1619-U100EA 77.8 EUR
70-1619-U100
Anti-KLRB1 Mouse Monoclonal Antibody [clone: HP-3G10]
Antibodies
The HP-3G10 antibody is specific for human CD161, also known as NKR-P1A, a type II transmembrane lectin-like receptor and member of the killer cell lectin-like receptor (KLR) family. CD161 exists as a homodimer which is prominently expressed on natural killer (NK) and NKT cells, where it is proposed to regulate the function of both cell types. CD161 is also found on T cell subsets, including T regulatory cells (Tregs), memory/effector CD4+ T cells, and CD8+ T cells. Th17 cells have been demonstrated to co-express CD161, as surface IL-17A+ cells are contained within the CD161+ fraction of CD4More Product Information
Order Now

SPECIFICATIONS

Antigen symbol KLRB1
Antigen name Killer cell lectin-like receptor subfamily B, member 1
Environmentally Preferable
Antigen synonyms NKRP1A|hNKR-P1A|NKR-P1A|NKR-P1|NKR|CLEC5B|CD161
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone HP-3G10
Reactivity Baboon,Rhesus monkey,Chimpanzee,Human
Host Mouse
Gene ID 3820
Isotype IgG1 kappa
Western blot Yes
Flow cytometry Yes
Cross adsorption none
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR