VWR® PerfectBlue Mini M, Gel System

Supplier: VWR Collection
700-0758EA 748 EUR
700-0758
VWR® PerfectBlue Mini M, Gel System
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
The PerfectBlue Mini M electrophoresis system is a very convenient size for a wide range of routine uses. Supplied with two combs, but with a wide range of standard and microtitre combs available, this system can be used for the separation of up to 36 samples. The system comes 'ready to use' with lid, power leads and a gel tray. Its high specification construction and embedded platinum electrodes provide high quality results time after time.

*Based on an agarose gel of approximately 5 mm thickness

Ordering information: System is supplied with 2 combs (10 and 14 teeth, 1,5 mm thickness) as standard.
Order Now

SPECIFICATIONS

Model Mini M
Environmentally Preferable
Gel size W×L 90×110 mm
Buffer volume 600 ml ml
Run time ≥45 h
Unit W×L×H 90×110×95 mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR