VWR® PerfectBlueMidi ExW Wide format gel system
Supplier: VWR Collection
700-0842EA 1350 EUR
700-0842
VWR® PerfectBlueMidi ExW Wide format gel system
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
With its extra wide design, the Midi ExW is ideal when there is a need to run lots of samples (up to 200) in just a short distance, for example, when genotyping. This format, along with its compact footprint, offers rapid run times without compromising on power. The result is a system for high throughput sample processing, generating crisp clear band data. As with all PerfectBlue gel systems, gels can be poured directly in the trays at temperatures up to +60 °C. With the Midi ExW, the trays can be walled with effective gasketed 'end gates' which are supplied in the system. In addition, gelMore Product Information
Order Now
SPECIFICATIONS
Model Midi ExW
Environmentally Preferable
Gel size W×L 230×140 mm
Sample capacity 25 – 200
Gel volume 160 ml
Buffer volume ~1400 ml
Run time ≥45 h
Unit W×L×H 260×290×100 mm