behrotest® DET supplementary set for Soxhlet extraction (30 ml), DE 30
Supplier: BEHR
B00218117
705-0026EA 2200 EUR
705-0026
behrotest® DET supplementary set for Soxhlet extraction (30 ml), DE 30
Extractors Soxhlet and Soxhlet-Type Extractors
This set includes the following equipment:
5×Sample vessels, 250 ml 65/250
5×Vessel –extractor connector M 2965
5×30 ml soxhlet extractor EZ 30
5×Reflux condenser for EZ 30 extractor RFK 30
Ordering information: The DET Basic unit (DBAS, 705-0023) is also required.