COD digestion workstations according to EPA, ISO or DIN
Supplier: BEHR
B00218408 B00218409 B00218405 B00218406 B00218407
705-0030EA 9690 EUR
705-0030 BEHRB00218409 705-0027 705-0028 705-0029
COD digestion workstations according to EPA, ISO or DIN
Digestion Systems Mineralisation Workstations
COD determination is a very important parameter for wastewater and sewage measurements. Behr workstations guarantee COD analysis in strict accordance with the following standards.
  • ISO 6060
  • DIN 38409 part 41 (COD >15 ppm)
  • DIN 38409 part 43 (COD abbreviated procedure)
  • DIN 38409 part 44 (COD 5 - 50 ppm)
Equipment configuration for simultaneous digestion of up to 6 or 12 COD samples.
Type 2 systems consist of the following components:
- Microprocessor-controlled TRS 300 time/temperature regulator with COD program
- Precision heating blockMore Product Information