Multiparameter photometer, MD 600
Supplier: Lovibond
214020
705-0920EA 1238.5 EUR
705-0920
Multiparameter photometer, MD 600
Colorimeters Multiparameter Colorimeters
The MD 600 is designed for the outdoor professional or on-site visits. It contains more than 120 pre-programmed methods.
  • Optional infrared data transfer module and connection cable with integrated electronics (RS232/RJ-45 plug)
  • Automatic wavelength selection
  • Waterproof
Delivery information: Supplied with carrying case, four AA/LR6 batteries, three round glass vials, adapter for 13 and 16 mm vials, plastic stirring rod, brush and screw driver.