Alkalinity standard solution, 25000 mg/L CaCO₃ (NIST)
Supplier: Hach
1427810
705-2018EA 67.1 EUR
705-2018
Alkalinity standard solution, 25000 mg/L CaCO₃ (NIST)
Standards Environmental Analysis Standards
Standard solution for accuracy checks in alkalinity determinations.
  • Simple accuracy checks
  • Minimise waste
  • Reliable results
Voluette and PourRite ampule standards are designed to work directly with standard additions accuracy checks in Hach procedures and photometer software. Use the Tensette pipet to add precise spikes of standard and check percent recovery, to confirm analytical system performance.