Swab with foam-covered cotton bud
Supplier: Texwipe
TX700B
710-0544EA 1270 EUR
710-0544
Swab with foam-covered cotton bud
Swabs Swabs for Cleaning
Swab with open-cell PU foam head. This surrounds a cotton bud core, which provides cushioned support and additional sorbency.
  • Highly absorbent
  • Low fibre and particle generation
  • Good chemical resistance
Used for cleaning surfaces and hard-to-reach areas.
Packaging: Bag-within-a-bag.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904