Large head swabs, Alpha®
Supplier: Texwipe
TX714A
710-0552EA 854 EUR
710-0552
Large head swabs, Alpha®
Swabs Swabs for Cleaning
This swab, for cleaning and validation sampling, features a 100% continuous filament polyester substrate that is heat-moulded (thermally bonded) onto the handle.
  • Sealed edges to reduce fibre release and eliminate the need for contaminating adhesives
  • Incorporates a full size, regular head on a rigid paddle
  • Polyester is soft, non abrasive and good for sensitive surfaces
Packaging: Cleanroom processed bag-within-a-bag.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904