VWR® Star-Blender® Digital Laboratory Blender
Supplier: VWR Collection
710-0958EA 3090 EUR
710-0958 710-1045 710-1044
VWR® Star-Blender® Digital Laboratory Blender
Homogenisers Laboratory Paddle Blenders
Sample volumes from 80 to 400 ml.
  • Digital display for easy setting of speed and time
  • Two reciprocating paddles
  • Polished AISI 304 stainless steel external and internal surfaces
  • Window in front door for observation of bagged samples during homogenisation
  • Tilting closure with removable front door to make operation and cleaning easier
Microbiological and pathogenic samples can be treated without risk to the operator or environment, by blending solid and semi-solid samples in sealed bags, eliminating bioaerosol production.