Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Tonbo Biosciences
72-8085-M001
72-8085-M001EA 226 EUR
72-8085-M001
Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Donkey anti-Sheep IgG (H&L) - Affinity Pure, HRP Conjugate, min x w/human, mouse, or rabbit IgG

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and light chain
Host: Donkey
Isotype:
Reactivity: Sheep
Order Now

SPECIFICATIONS

Antigen symbol IgG
Environmentally Preferable
Antigen name Immunoglobulin G
Antibody type Secondary
Clonality Polyclonal
Conjugation HRP (Horseradish Peroxidase)
Reactivity Sheep
Host Donkey
Isotype
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Epitope Heavy and light chain
Cross adsorption Yes
Storage temperature Store freeze-dried powder at 2-8 °C. Once rehydrated and in solution store at 2-8 °C.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR