Staining jars acc. to Coplin, WHEATON®
Supplier: DWK Life Sciences
900520
720-0707EA 263 EUR
720-0707
Staining jars acc. to Coplin, WHEATON®
Jars Staining Jars
Soda-lime glass.
  • Slides extend above the opening
  • Easy manipulation without using forceps
  • Stable rectangular base
This staining jar has the capacity for up to 10 standard slides of 76×26 mm with white PP screw cap.