Transport swabs, Transwab®

Supplier: MEDICAL WIRE EQUIPMENT
MW170 MW171 MW172P MW173C MW173P MW172C
720-0138EA 105 EUR
720-0138 720-0139 720-0140 720-2201 720-2202 MEDWMW172C
Transport swabs, Transwab®
Swabs Transport Swabs
Each transport tube contains Amies medium preferred because the inorganic buffer ensures maintenance without overgrowth.

  • Excellent recovery of both aerobic and anaerobic bacteria
  • High release of micro-organisms onto plates or into broth
  • Safety cap includes double seal, inside and outside tube
  • Colour coded caps for ease of selection
  • Pre-labelled transport tube

Certifications: MDD, IVD, and CLSI M40.

Packaging: Easy peel pack.

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR