IP ActivFlo Biopsy III cassettes, taped
Supplier: LEICA HISTOLOGY
39LC-625-1-L 39LC-625-10-L 39LC-625-11-L 39LC-625-2-L 39LC-625-3-L 39LC-625-4-L 39LC-625-5-L 39LC-625-6-L 39LC-625-7-L 39LC-625-8-L 39LC-625-9-L
720-2364EA 352 EUR
720-2364 720-2653 720-2655 720-2657 720-2659 720-2661 720-2663 720-2665 720-2667 720-2669 720-2671
IP ActivFlo Biopsy III cassettes, taped
Embedding Cassettes Biopsy Processing Cassettes
IP ActivFlo Biopsy III cassettes feature a patented 'Fluidic Flow' pore design for maximum reagent exchange. With a 0,26 mm pore size, the cassettes can secure the smallest specimens, eliminating the need for biopsy bags and sponges. The cassettes also feature sloped walls, creating a basket for small tissue management. These two piece cassettes with hinged lid have been designed for the Leica IP C Cassette Printer, with <1% printing error rate.
  • Available in taped packaging for easy printer loading
  • Eleven colours available