Heat sealing foil, ClearASeal Peel™
Supplier: iST Scientific Ltd
IST-101-115LR IST-101-078LR IST-101-078LS IST-101-078SR IST-101-078SS IST-101-078TR IST-101-078TS IST-101-115SR IST-101-115TR
731-0477EA 558 EUR
731-0477 731-0475 731-0479 731-0476 731-0480 731-0481 731-0483 731-0478 731-0482
Heat sealing foil, ClearASeal Peel™
Seals Microplate Seals
Heat sealing offers a 100% effective method for plate sealing for a complete seal integrity, as well as being quick and cost effective. The ClearASeal Peel is an optically clear laminate film forming a peelable seal to polypropylene, polyethylene, polystyrene, polycarbonate and cyclic olefin copolymer (COC) plates
  • Moderate solvent resistance, can be utilized for short term compound storage at room temperature
  • Available as sheets, for use with manual and semi-automated sealers, or in multiple roll formats compatible with specified automated heat sealers