Whatman™ Protran® BA85 & BA83 NC Western Blotting Membranes, Nitrocellulose
Supplier: Whatman products (Cytiva)
10401116 10402506 10402525 10402578 10401316 10401403
732-4000EA 322 EUR
732-4000 732-4168 732-4169 732-0347 732-4012 WHAT10401403
Whatman™ Protran® BA85 & BA83 NC Western Blotting Membranes, Nitrocellulose
Transfer Membranes
Protran® membranes are manufactured from 100% pure nitrocellulose to ensure the highest binding capacity possible. They are compatible with a variety of detection methods, including isotopic, chemiluminescent, colorimetric and fluorescent. No methanol pre-wetting step is required, making this the membrane of choice for proteins which prefer aqueous environments. Prior to transfer the membrane is simply wetted with water, and then placed in the transfer buffer. No other pre treatment steps are necessary.
  • High binding, low background, without the need for stringent washing conditions