Pipette tips, gel loading, HydroLogix™
Supplier: Thermo Fisher Scientific
3671-11 3631 3691-11
732-2339EA 0 EUR
732-2339 732-2256 MLBP3691-11
Pipette tips, gel loading, HydroLogix™
Pipette Tips
Designed for loading vertical electrophoresis gels.
  • Choice of round or flat tip
  • Available in volumes 1 to 10 µl or 2 to 200 µl
  • Intended for loading gels of three thicknesses to suit specific applications