Microplates, high content imaging glass bottom, Corning®
Supplier: Corning
4580 4582 4584
732-2493EA 323.26 EUR
732-2493 BDAA4582 BDAA4584
Microplates, high content imaging glass bottom, Corning®
Microplates HTS Microplates
High optical quality glass bottom, black microplates are ideal for performing high content cell based assays using imaging systems. The glass bottom provides a flat and optically clear surface that reduces autofocus time, increases throughput and is ideal for cell growth.
  • High optical quality and scratch-resistant glass
  • Glass bottom thickness of 200 µm is well suited for imaging microscopy
  • Well bottom flatness <50 µm to ensure planarity for imaging devices
  • Low background fluorescence and minimal crosstalk to provide the highest possible optical quality