VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
734-2328EA 367 EUR
734-2328 734-2323 734-2324 734-2325 734-2326 734-2327 732-2904 732-2906 732-2907 732-2905 734-2777 734-2778 734-2779 734-2780 734-2781 734-2782 734-2798 734-2799 734-2800 734-2801 734-2802 734-2803 734-2888
VWR® Multiwell Cell Culture Plates
Microplates Cell Culture Plates
VWR® multiwell cell culture plates are available with a choice of surface treatments.
  • Uniform well volume ensures an equal growth surface area
  • Well surface is smooth and free from striations to maximise usable growth area
  • Raised rims on wells with uniform rings on the lid to reduce evaporation
  • Single-position lid reduces the risk of cross-contamination and handling mistakes
  • Wells are labelled with alphanumeric code for easy identification
  • Suitable for use with all common instruments and automation
  • Sterilised by electron beam irradiation
  • Free from DNase and
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001/Certificate No.: 04923E00243R0M