Peqlab, PCR Plates, 96-Well, Non-Skirted
Supplier: VWR Peqlab
732-3214EA 154 EUR
732-3214 732-2879
Peqlab, PCR Plates, 96-Well, Non-Skirted
Microplates PCR Plates
PP, non-skirted. Standard 96-well PCR plates, suitable for a wide range of 0,2 ml thermal cycler blocks.
  • Effective design with overhanging edges for easy handling
  • Cut corner (H1)
  • Can be sealed using Thermo-Mats™, adhesive films and foils, heat seals and flat or domed cap strips
  • White plates for Real-Time PCR
  • Certified free from DNase, RNase and human DNA
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification IRECS; ISO 14001 (Certificate Registration No. EMS 752603)