Electroporation system, twin-wave, Gemini X² and SC

Supplier: BTX
45-2044 45-2040 45-2041 45-2042 45-2043
732-3775EA 20810 EUR
732-3775 732-3771 732-3772 732-3773 732-3774
Electroporation system, twin-wave, Gemini X² and SC
Electroporation Systems Electroporators
Gemini twin-wave electroporators are flexible systems allowing both square wave and exponential decay wave electroporation in a single unit. These waveform combinations enable researchers to easily and efficiently electroporate eukaryotic and prokaryotic cells in cuvettes or plates with one easy to use set-up. Ideal for all the electroporation needs, including CRISPR, in vivo, in vitro, in ovo and more.

  • Universal electroporation – transfects cells in vitro, in vivo, in ovo and adherent forms
  • Multiple pulsing capabilities in both square and exponential
More Product Information
Order Now

SPECIFICATIONS

W×D×H 286×216×324 mm 286×216×324 mm 286×216×324 mm
Model Gemini X2 HT Gemini X2 Gemini SC
Environmentally Preferable
Interfaces Simple user interface Simple user interface Simple user interface
Charging time <7 s <7 s <7 s
Voltage range High Voltage Mode / 5 V resolution 505 - 3000 V 505 - 3000 V 505 - 3000 V
Voltage range High Voltage Mode / 10 V resolution 510 - 3000 V
Voltage range Low Voltage Mode / 1 V resolution 5 - 500 V 5 - 500 V
Multiple pulsing 1 - 10 pulses per sample (up to 120 total) 1 - 99 pulses per sample 1 - 10 pulses per sample (low voltage mode);
1 - 2 pulses per sample (high voltage mode)
Pulse interval 0,1 - 10 s; 5 - 30 s 0,1 - 10 s; 5 - 30 s 0,1 - 10 s
Pulse length range, HV mode/1 μs resolution 10 - 600 μs 10 - 600 μs
Pulse length range, LV mode/1 μs resolution 10 - 999 μs 10 - 999 μs
Pulse length range, LV mode/1 ms resolution 1 - 999 ms 1 - 999 ms 10 - 100 ms
Capacitance in high voltage mode 85 µF 85 µF 85 µF
Capacitance in low voltage mode 3775 µF 3775 µF 3775 µF
Display Touch screen Touch screen Touch screen
Weight 7,3 kg 7,3 kg 7,3 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR