VWR®, PCR Workstation
Supplier: VWR Collection
732-2541EA 3220 EUR
732-2541 732-2542 732-2840 732-3900
VWR®, PCR Workstation
Fume Hoods and Enclosures PCR Workstation Enclosures
The VWR® PCR Workstation is a PCR enclosure for PCR sample preparation and other sensitive protocols. It offers dual decontamination action by UV inactivation of airborne and surface-bound contaminants and, therefore, represents an ideal environment for PCR set up. Stainless steel housing, front and side panels made of 8 mm Makrolon® protects from UV irradiation.
  • Active decontamination of work surface during non working time by UV irradiation
  • Additional inactivation of aerosol-bound contaminants by shielded UV Air Recirculator during operation
  • Contaminant