dNTP mix
Supplier: Quantabio
95062-01K
733-1271EA 481 EUR
733-1271
dNTP mix
Nucleic Acid Reagents Nucleotides
The 10 mM dNTP mix is a solution of high purity deoxyribonucleoside triphosphates that has been functionally qualified for real-time quantitative PCR (qPCR). It is suitable for use in conventional end-point PCR, real time qPCR, first-strand cDNA synthesis, as well as other applications that require dNTP as substrate. The use of qPCR certified dNTPs is a simple but important measure to ensure consistent assay performance.