Automated cell counter, EVE™ Plus
Supplier: NANOENTEK
EVE-MC2-EU EVE-MC2-UK
734-3333EA 4900 EUR
734-3333 NANOEVE-MC2-UK
Automated cell counter, EVE™ Plus
Cell Counters and Flow Cytometers Automated Cell Counters
EVE™ Plus is a compact image-based automated cell counter that uses disposable slides to most accurately determine eukaryotic cell titers in less than one second. In addition, when applying trypan blue staining, the system will discriminate between live from dead cells precisely to provide reliable data on cell viability. Compared to counting cells manually using classical haemocytometers, the EVE™ PLUS automated cell counter offers three main benefits.
  • Reproducible, objective data, free from human bias/error
  • Quick and lean workflow saves times and
More Product Information
Order Now
SPECIFICATIONS
Concentration 1×10⁴ - 2×10⁷ cells/ml
Environmentally Preferable
Cell size 5 - 60 µm
Sample volume 10 µl
Focus Manual or automatic
Measurement time 1 s/test (manual focus), <10 s/test (auto focus)