Crystallisation plates, CrystalStar™, 96-well CrystalQuick™ and CrystalDrop™
Supplier: Greiner Bio-One
609101_40 609120_40 609171_80 609820_40 609801_40 609871_80 609180_80
737-3501EA 675 EUR
737-3501 736-0222 737-3502 737-3504 737-3503 737-3505 737-3525
Crystallisation plates, CrystalStar™, 96-well CrystalQuick™ and CrystalDrop™
Microplates Crystallisation Plates
PS, except LBR plates (polyolefin). The CrystalStar™ product range is a family of crystallisation plates and accessories designed specifically for high throughput crystallisation.
  • All CrystalStar™ plates, with the exception of Terasaki plates, conform to SBS standard
  • Plates with hydrophobic surface (CrystalQuick™ Plus) are suited for nanolitre crystallisation of membrane proteins
  • LBR (low birefringence) plates are specifically designed for use with polarised light
These plates are designed for sitting drop applications. Each of the 96 reservoirsMore Product Information