Fast wet transfer system for mini gels, eBlot® L1
Supplier: Genscript
L00686 L00724 L00726 L00727
786-0002EA 8210 EUR
786-0002 786-0003 786-0004 786-0005
Fast wet transfer system for mini gels, eBlot® L1
Blotting Apparatus
eBlot® L1 wet protein transfer system is a highly efficient western blotting transfer system that uses a novel technology (patent pending) developed by GenScript. eBlot® L1 combines the high reproducibility and efficiency of the traditional wet transfer with the speed and convenience of other fast transfer systems. The device allows efficient blot transfer of small, medium and large molecular weight proteins within 9 to 17 minutes.
  • Benefits of eBlot® L1
  • Comprehensive: Outstanding performance for small, medium and large proteins
  • Convenience: Simultaneously