Dot blot system, Enhancer™
Supplier: G-Biosciences
786-163
786-163EA 2350 EUR
786-163
Dot blot system, Enhancer™
Blotting Apparatus
Enhancer™ dot blot system allows for the rapid application of up to 384 protein or nucleic acid samples on to a 12×9 cm membrane for subsequent analysis. Samples are applied to a nitrocellulose or nylon membrane by capillary action, resulting in concentration of sample due to minimal diffusion.
  • Increase dot blot sensitivity by 10-fold
  • Apply up to 384 protein or nucleic acid samples
  • No cross-contamination risk
  • No expensive electronic equipment required