Adenine deaminase
Supplier: G Bioscienes
786-428
786-428EA 724 EUR
786-428
Adenine deaminase
Protein Extraction Protein Extraction Reagents
Adenine deaminase converts adenine to hypoxanthine. It is a highly purified recombinant enzyme protein supplied at a 1 µM concentration.