J.T.Baker® BAKERBOND Speedisk® Hydrophobic SC-DVB (SO3), SPE Columns
Supplier: Avantor
8196-02EA 0 EUR
8196-02
J.T.Baker® BAKERBOND Speedisk® Hydrophobic SC-DVB (SO3), SPE Columns
Chromatography Columns
These columns feature BAKERBOND® DVB polymer resins, which are the product of ultra-clean polymer microparticle technology. These resin particles have a large surface area, are highly rigid, are stable over pH range 1 to 14, are water-wettable and are not impacted by sorbent drying. These columns are recommended when advanced detection methods are used.
  • Improve sample preparation recovery
  • Polymer based
General applications: Ionic, basic analytes.
These SPE columns can be used in mixed mode.