1-Propanol 99,8, HiPerSolv CHROMANORM® HPLC
Supplier: VWR Chemicals
Danger
Synonyms: Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane
Filter 0,2 um_Nitrogen_filled
83635.320EA 277 EUR
83635.320
1-Propanol 99,8, HiPerSolv CHROMANORM® HPLC
1-Propanol
Formula: H₃CCH₂CH₂OH
MW: 60.1 g/mol
Boiling Pt: 97
Melting Pt: –127 °C
Density: 0.8053 g/cm³ (20 °C. DIN 51757)
Flash Pt: 15
Storage Temperature: Ambient
MDL Number: MFCD00002941
CAS Number: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3 II
Merck Index: 13,07934
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay (GC) Min. 99.80 %
Acidity Max. 0.0005 %
Alkalinity Max. 0.0002 %
Evaporation residue (100°C) Max. 0.0010 %
Water Max. 0.0500 %
Transmittance (230 nm) Min. 50.0 %
Transmittance (240 nm) Min. 80.0 %
Transmittance (270 nm) 98.0 %
Conforms to BDH 15303 Passes test